Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Optineurin (OPTN) Monoclonal Antibody (OTI7H2), TrueMAB™, OriGene
SDP

Catalog No. 502543797
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3797 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3797 Supplier OriGene Supplier No. CF812132
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Optineurin (OPTN) Monoclonal antibody specifically detects Optineurin (OPTN) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Optineurin (OPTN)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7H2
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DBN1
Gene Accession No. Q07266, Q16643, Q9QXS6
Gene Alias 14.7K-interacting protein 2; 4930441O07Rik; Ag9 C5; ALS12; E3-14.7K-interacting protein; FIP 2; FIP2; FIP-2; FIP-2-like protein; GLC1E; HIP 7; HIP7; HIP-7; Huntingtin interacting protein L; huntingtin yeast partner L; Huntingtin-interacting protein 7; huntingtin-interacting protein L; HYPL; NEMO-related protein; NRP; Optic neuropathy-inducing protein; optineurin; Optn; TFIIIA IntP; TFIIIA-IntP; transcription factor IIIA-interacting protein; transcrption factor IIIA-interacting protein; tumor necrosis factor alpha-inducible cellular protein containing leucine zipper domains; Uban
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-240 of human DBN1 (NP_004386) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1627, 56320, 81653
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.