Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTN Mouse anti-Human, Biotin, Clone: 6C1H4, Proteintech
SDP

Catalog No. 501737227 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7227 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7227 Supplier Proteintech Group Inc Supplier No. BIOTIN60293100UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is conjugated with Biotin.

This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14.7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.

Specifications

Antigen OPTN
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6C1H4
Concentration 0.73 mg/mL
Conjugate Biotin
Formulation PBS with 50% glycerol and no preservative; pH 7.3
Gene OPTN
Gene Accession No. Q96CV9
Gene Alias FIP 2, optineurin, OPTN
Gene Symbols OPTN
Host Species Mouse
Immunogen OPTN Fusion Protein Ag1272
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10133
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.