Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR10A2 Polyclonal Antibody
OR10A2 Polyclonal Antibody
Supplier: Thermo Scientific PA5144366
Description
OR10A2 Polyclonal Antibody for ICC/IF, IHC (P), ELISA

Specifications
OR10A2 | |
Polyclonal | |
Unconjugated | |
OR10A2 | |
HP4; hP4 olfactory receptor; olfactory receptor 10A2; olfactory receptor family 10 subfamily A member 2; olfactory receptor OR11-82 pseudogene; olfactory receptor OR11-86; olfactory receptor, family 10, subfamily A, member 2 pseudogene; OR10A2; OR10A2P; OR11-82; OR11-86; OST363 | |
Rabbit | |
Protein G | |
RUO | |
341276 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry | |
3.13 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q9H208 | |
OR10A2 | |
Peptide sequence from Human Olfactory receptor 10A2 protein (247-262AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction