Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR10H5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548396
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548396 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548396 Supplier Invitrogen™ Supplier No. PA548396
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR10H5 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR10H5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR10H5
Gene Accession No. Q8NGA6
Gene Alias Olfactory receptor 10H5; olfactory receptor family 10 subfamily H member 5; olfactory receptor OR19-25; Olfactory receptor OR19-26; olfactory receptor, family 10, subfamily H, member 5; OR10H5; OR19-25; OR19-26
Gene Symbols OR10H5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 245-273 amino acids from the C-terminal region of human OR10H5.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 284433
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.