Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR10K1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563660 Supplier Invitrogen™ Supplier No. PA563660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TSLVFHLPFH SSNQLHHFFC DI Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 95%.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR10K1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR10K1
Gene Accession No. Q8NGX5
Gene Alias Olfactory receptor 10K1; Olfactory receptor OR1-6; OR10K1
Gene Symbols OR10K1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR10K1/OR10K2. Recombinant protein control fragment (Product #RP-101773).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391109
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.