Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR10K1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144367
Description
OR10K1 Polyclonal Antibody for ICC/IF, ELISA
Odorant receptor.
Specifications
OR10K1 | |
Polyclonal | |
Unconjugated | |
OR10K1 | |
Olfactory receptor 10K1; Olfactory receptor OR1-6; OR10K1 | |
Rabbit | |
Protein G | |
RUO | |
391109 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunocytochemistry | |
4.47 mg/mL | |
PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
Q8NGX5 | |
OR10K1 | |
Peptide sequence from Human Olfactory receptor 10K1 protein (259-272AA). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction