Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR10Q1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100971
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100971 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100971 Supplier Invitrogen™ Supplier No. PA5100971
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR10Q1.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR10Q1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR10Q1
Gene Accession No. Q8NGQ4
Gene Alias olfactory receptor 10Q1; olfactory receptor family 10 subfamily Q member 1; olfactory receptor OR11-233; olfactory receptor, family 10, subfamily Q, member 1; OR10Q1; OR11-233
Gene Symbols OR10Q1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR10Q1(Accession Q8NGQ4), corresponding to amino acid residues L250-K300.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219960
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.