Learn More
Invitrogen™ OR10S1 Polyclonal Antibody

Description
Immunogen sequence: MTSRSVCEKM TMTTENPNQT VVSHFFLEGL RYTAKHSSL Highest antigen sequence identity to the following orthologs: Mouse - 41%, Rat - 41%.
Specifications
Specifications
| Antigen | OR10S1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | OR10S1 |
| Gene Accession No. | Q8NGN2 |
| Gene Alias | Olfactory receptor 10S1; olfactory receptor family 10 subfamily S member 1; olfactory receptor OR11-279; olfactory receptor, family 10, subfamily S, member 1; OR10S1; OR11-279 |
| Gene Symbols | OR10S1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.