Learn More
Invitrogen™ OR10W1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total OR10W1.
Specifications
Specifications
| Antigen | OR10W1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | OR10W1 |
| Gene Accession No. | Q8NGF6 |
| Gene Alias | Olfactory receptor 10W1; olfactory receptor family 10 subfamily W member 1; olfactory receptor OR11-236; olfactory receptor, family 10, subfamily W, member 1 pseudogene; OR10W1; OR10W1P; OR10W1Q; OR11-236; UNQ6469; UNQ6469/PRO34070 |
| Gene Symbols | OR10W1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.