Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR11A1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | OR11A1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
OR11A1 Polyclonal specifically detects OR11A1 in Human samples. It is validated for Western Blot.Specifications
OR11A1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
dJ994E9.6, family 11, subfamily A, member 2, Hs6M1-18, olfactory receptor 11A1, Olfactory receptor 11A2, Olfactory receptor OR6-30, olfactory receptor, family 11, subfamily A, member 1, OR11A2 | |
The immunogen is a synthetic peptide directed towards the C terminal region of human OR11A1 (NP_039225.1). Peptide sequence YVAPSAVHSQLLSKVFSLLYTVVTPLFNPVIYTMRNKEVHQALRKILCIK | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
26531 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title