Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR13C2/OR13C9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568538
Description
This target displays homology in the following species: Cow: 79%; Dog: 93%; Horse: 86%; Human: 100%; Pig: 79%; Rat: 77%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
OR13C2/OR13C9 | |
Polyclonal | |
Unconjugated | |
OR13C2 | |
cOR13C9; olfactory receptor 13C2; Olfactory receptor 13C9; olfactory receptor family 13 subfamily C member 2; olfactory receptor family 13 subfamily C member 2B; olfactory receptor family 13 subfamily C member 9; olfactory receptor OR9-12; Olfactory receptor OR9-13; olfactory receptor, family 13, subfamily C, member 2; olfactory receptor, family 13, subfamily C, member 9; OR13C2; OR13C2B; OR13C9; OR37K; OR37L; OR9-13 | |
Rabbit | |
Affinity chromatography | |
RUO | |
286362, 392376 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NGS9, Q8NGT0 | |
OR13C2, OR13C9 | |
synthetic peptide directed towards the middle region of human OR13C2/OR13C9. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction