Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR13C9 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | OR13C9 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
OR13C9 Polyclonal specifically detects OR13C9 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
OR13C9 | |
Polyclonal | |
Purified | |
RUO | |
olfactory receptor, family 13, subfamily C, member 9, OR37L | |
OR13C9 | |
IgG | |
Protein A purified |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
NP_001001956 | |
286362 | |
Synthetic peptide directed towards the middle region of human OR13C9. Peptide sequence IFYGTILFMYMKPKSKETLNSDDLDATDKIISMFYGVMTPMMNPLIYSLR. | |
Primary | |
36 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title