Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR13D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563208
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563208 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563208 Supplier Invitrogen™ Supplier No. PA563208
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISIYLNHVLF YTTQQAGDLE HMETRNYSAM TEFFLV Highest antigen sequence identity to the following orthologs: Mouse - 33%, Rat - 31%.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR13D1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR13D1
Gene Accession No. Q8NGV5
Gene Alias olfactory receptor 13D1; olfactory receptor family 13 subfamily D member 1; olfactory receptor OR9-15; olfactory receptor, family 13, subfamily D, member 1; OR13D1; OR9-15
Gene Symbols OR13D1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR13D1. Recombinant protein control fragment (Product #RP-100768).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286365
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.