Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR14C36 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548627
Description
OR14C36 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
OR14C36 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NHC7 | |
OR14C36 | |
KLH conjugated synthetic peptide between 284-312 amino acids from the C-terminal region of human OR14C36. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR14C36 | |
Olfactory receptor 14C36; olfactory receptor 5BF1; olfactory receptor family 14 subfamily C member 36; olfactory receptor OR1-59; olfactory receptor, family 14, subfamily C, member 36; olfactory receptor, family 5, subfamily BF, member 1; OR14C36; OR5BF1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
127066 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction