Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR14I1 Rabbit anti-Human, Polyclonal, FabGennix

Catalog No. 502208041
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-8041 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-8041 Supplier Fabgennix Inc Supplier No. OR14I1101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR14I1 Polyclonal antibody specifically detects OR14I1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen OR14I1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene OR14I1
Gene Accession No. A6ND48
Gene Alias olfactory receptor 14I1; Olfactory receptor 5BU1; olfactory receptor family 14 subfamily I member 1; olfactory receptor, family 14, subfamily I, member 1; olfactory receptor, family 5, subfamily BU, member 1 pseudogene; OR14I1; OR5BU1; OR5BU1P
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1-50 on human Olfactory Receptor, family 14, subfamily I, member 1 protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401994
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.