Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR1A2 Polyclonal Antibody
OR1A2 Polyclonal Antibody
Supplier: Thermo Scientific PA570835
Description
OR1A2 Polyclonal Antibody for Western Blot

Specifications
OR1A2 | |
Polyclonal | |
Unconjugated | |
OR1A2 | |
Olfactory receptor 17-6; olfactory receptor 1A2; olfactory receptor family 1 subfamily A member 2; olfactory receptor OR17-10; olfactory receptor, family 1, subfamily A, member 2; OR17-6; OR1A2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26189 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y585 | |
OR1A2 | |
synthetic peptide directed towards the C-terminal region of human OR1A2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction