Learn More
Invitrogen™ OR1D2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total OR1D2.
Specifications
Specifications
| Antigen | OR1D2 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | OR1D2 |
| Gene Accession No. | P34982 |
| Gene Alias | olfactory receptor 17-4; Olfactory receptor 1D2; olfactory receptor family 1 subfamily D member 2; olfactory receptor OR17-6; olfactory receptor, family 1, subfamily D, member 2; olfactory receptor-like protein HGMP07E; OLFR1; OR17-4; OR1D2 |
| Gene Symbols | OR1D2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.