Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR1D4/OR1D5 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103224
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103224 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103224 Supplier Thermo Scientific Supplier No. PA5103224

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR1D4/5.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR1D4/OR1D5
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR1D4
Gene Accession No. P47884, P58170
Gene Alias C17orf2; Olfactory receptor 17-31; olfactory receptor 1D5; olfactory receptor family 1 subfamily D member 4 (gene/pseudogene); olfactory receptor family 1 subfamily D member 5; olfactory receptor OR17-2; olfactory receptor, family 1, subfamily D, member 5; OR17-2; OR17-30; OR17-31; OR1D4; OR1D5
Gene Symbols OR1D4, OR1D5
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR1D5(Accession P58170), corresponding to amino acid residues I206-A256.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 653166, 8386
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.