Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR1F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548960
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548960 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548960 Supplier Invitrogen™ Supplier No. PA548960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR1F1 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR1F1
Applications Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR1F1
Gene Accession No. O43749
Gene Alias Olfactory receptor 16-35; Olfactory receptor 1F1; Olfactory receptor 1F10; Olfactory receptor 1F4; Olfactory receptor 1F5; Olfactory receptor 1F6; Olfactory receptor 1F7; Olfactory receptor 1F8; Olfactory receptor 1F9; olfactory receptor family 1 subfamily F member 1; Olfactory receptor OR16-4; olfactory receptor, family 1, subfamily F, member 1; olfactory receptor, family 1, subfamily F, member 10; olfactory receptor, family 1, subfamily F, member 13 pseudogene; olfactory receptor, family 1, subfamily F, member 4; olfactory receptor, family 1, subfamily F, member 5; olfactory receptor, family 1, subfamily F, member 6; olfactory receptor, family 1, subfamily F, member 7; olfactory receptor, family 1, subfamily F, member 9; OLFMF; OR16-35; OR16-36; OR16-37; OR16-88; OR16-89; OR16-90; OR1F1; OR1F10; OR1F13P; OR1F4; OR1F5; OR1F6; OR1F7; OR1F8; OR1F9; OR3-145; ORL1023
Gene Symbols OR1F1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 275-303 amino acids from the C-terminal region of human OR1F1.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4992
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.