Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR1L3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563623
Description
Immunogen sequence: QKLINKIKSQ MSRFSTKTNK Highest antigen sequence identity to the following orthologs: Mouse - 55%, Rat - 55%.
Odorant receptor (Potential).
Specifications
OR1L3 | |
Polyclonal | |
Unconjugated | |
OR1L3 | |
Olfactory receptor 1L3; olfactory receptor 9-D; olfactory receptor family 1 subfamily L member 3; olfactory receptor OR9-28; olfactory receptor, family 1, subfamily L, member 3; OR1L3; OR9-28; OR9-D | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NH93 | |
OR1L3 | |
Recombinant protein corresponding to Human OR1L3. Recombinant protein control fragment (Product #RP-100538). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction