Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR1L3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100979 Supplier Invitrogen™ Supplier No. PA5100979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR1L3.

Odorant receptor (Potential).
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR1L3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR1L3
Gene Accession No. Q8NH93
Gene Alias Olfactory receptor 1L3; olfactory receptor 9-D; olfactory receptor family 1 subfamily L member 3; olfactory receptor OR9-28; olfactory receptor, family 1, subfamily L, member 3; OR1L3; OR9-28; OR9-D
Gene Symbols OR1L3
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR1L3(Accession Q8NH93), corresponding to amino acid residues Y289-P324.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26735
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.