Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR1N1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA571849
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
OR1N1 | |
Polyclonal | |
Unconjugated | |
OR1N1 | |
Olfactory receptor 1N1; Olfactory receptor 1-26; OR1-26; Olfactory receptor 1N3; Olfactory receptor OR9-22; OR1N1; OR1N3 | |
Rabbit | |
Protein A | |
RUO | |
138883 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.91 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NGS0 | |
OR1N1 | |
KLH conjugated synthetic peptide between 277-306 amino acids from the C-terminal region of human OR1N1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction