Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR1S1/OR1S2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561927
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561927 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561927 Supplier Invitrogen™ Supplier No. PA561927
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSSFLQIGRN MHQGNQTTIT EFILL Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 52%.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR1S1/OR1S2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR1S1
Gene Accession No. Q8NGQ3, Q8NH92
Gene Alias Olfactory receptor 1S1; olfactory receptor 1S2; olfactory receptor family 1 subfamily S member 1 (gene/pseudogene); olfactory receptor family 1 subfamily S member 2; olfactory receptor OR11-231; Olfactory receptor OR11-232; olfactory receptor, family 1, subfamily S, member 1; olfactory receptor, family 1, subfamily S, member 2; OR11-231; OR11-232; OR1S1; OR1S2; OST034
Gene Symbols OR1S1, OR1S2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR1S1/OR1S2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219958, 219959
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.