Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549218
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549218 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549218 Supplier Thermo Scientific Supplier No. PA549218
Add to Cart
Add to Cart

OR2A12 Polyclonal Antibody

OR2A12 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2A12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR2A12
Gene Accession No. Q8NGT7
Gene Alias Olfactory receptor 2A12; olfactory receptor family 2 subfamily A member 12; olfactory receptor OR7-10; olfactory receptor, family 2, subfamily A, member 12 pseudogene; olfactory receptor, family 2, subfamily A, member 16 pseudogene; OR2A12; OR2A12P; OR2A16P
Gene Symbols OR2A12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 243-271 amino acids from the C-terminal region of human OR2A12.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 346525
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.