Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144368
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144368 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144368 Supplier Invitrogen™ Supplier No. PA5144368
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR2A7 Polyclonal Antibody for ICC/IF, ELISA

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2A7
Applications ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 1.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene OR2A7
Gene Accession No. Q96R45
Gene Alias Olfactory receptor 2A7; Olfactory receptor OR7-18; OR2A7
Gene Symbols OR2A7
Host Species Rabbit
Immunogen Peptide sequence from Human Olfactory receptor 2A7 protein (258-273AA).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401427
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.