Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR51J1 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5100937
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100937 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100937 Supplier Thermo Scientific Supplier No. PA5100937

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HOR5'Beta8.

OR51J1 is an odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR51J1
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR51J1
Gene Accession No. Q9H342
Gene Alias OR51J2
Gene Symbols OR51J1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR51J1(Accession Q9H342), corresponding to amino acid residues G210-L260.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79470
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.