Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR2AG1/OR2AG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538166
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
OR2AG1/OR2AG2 | |
Polyclonal | |
Unconjugated | |
OR2AG1 | |
OR2AG2P, Olfactory receptor 2AG2, OR11-79, OR2AG3, hT3 olfactory receptor, olfactory receptor 2AG1, olfactory receptor 2AG3, olfactory receptor OR11-79, olfactory receptor, family 2, subfamily AG, member 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
144125, 338755 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A6NM03, Q9H205 | |
OR2AG1, OR2AG2 | |
A synthetic peptide derived from the internal region of human OR2AG1/2AG2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction