Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2AT4 Antibody, Novus Biologicals™
SDP

Catalog No. p-7108354 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP159861 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP159861 Supplier Novus Biologicals Supplier No. NBP159861
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR2AT4 Polyclonal specifically detects OR2AT4 in Human samples. It is validated for Western Blot.

Specifications

Antigen OR2AT4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. A6NND4
Gene Alias olfactory receptor 2AT4, Olfactory receptor OR11-265, olfactory receptor OR11-265 pseudogene, olfactory receptor, family 2, subfamily AT, member 4, OR11-265
Gene Symbols OR2AT4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to OR2AT4(olfactory receptor, family 2, subfamily AT, member 4) Antibody(against the N terminal of OR2AT4. Peptide sequence MDATACNESVDGSPVFYLLGIPSLPETFFLPVFFIFLLFYLLILMGNALI.
Molecular Weight of Antigen 35 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341152
Test Specificity Expected identity based on immunogen sequence: Equine 77%, Mouse 85%, Rabbit: 77%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Pig, Bovine, Equine, Guinea Pig, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.