Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR2F2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5102285
Description
Antibody detects endogenous levels of total OR2F2.
Odorant receptor.
Specifications
OR2F2 | |
Polyclonal | |
Unconjugated | |
OR2F2 | |
Olfactory receptor 2F2; olfactory receptor 7-1; olfactory receptor family 2 subfamily F member 2; olfactory receptor OR7-6; olfactory receptor, family 2, subfamily F, member 2; OR2F2; OR7-1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
135948 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O95006 | |
OR2F2 | |
A synthesized peptide derived from human OR2F2(Accession O95006), corresponding to amino acid residues L101-S151. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction