Learn More
Invitrogen™ OR2F2 Polyclonal Antibody

Description
OR2F2 Polyclonal Antibody for Western Blot, Flow
Specifications
Specifications
| Antigen | OR2F2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | OR2F2 |
| Gene Accession No. | O95006 |
| Gene Alias | Olfactory receptor 2F2; olfactory receptor 7-1; olfactory receptor family 2 subfamily F member 2; olfactory receptor OR7-6; olfactory receptor, family 2, subfamily F, member 2; OR2F2; OR7-1 |
| Gene Symbols | OR2F2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.