Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR2G6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548738
Description
OR2G6 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
OR2G6 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q5TZ20 | |
OR2G6 | |
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human OR2G6. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR2G6 | |
Olfactory receptor 2G6; olfactory receptor family 2 subfamily G member 6; olfactory receptor, family 2, subfamily G, member 6; OR2G6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
391211 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction