Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102175
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102175 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102175 Supplier Invitrogen™ Supplier No. PA5102175
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR2H2.

OR2H2 is a odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2H2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR2H2
Gene Accession No. O95918
Gene Alias dJ271M21.2; FAT11; hs6M1-12; Olfactory receptor 2; olfactory receptor 2H2; Olfactory receptor 2H3; olfactory receptor family 2 subfamily H member 2; olfactory receptor OR6-36; olfactory receptor, family 2, subfamily H, member 2; olfactory receptor, family 2, subfamily H, member 3; olfactory receptor-like protein FAT11; OLFR2; OLFR42B; OR2H2; OR2H3
Gene Symbols OR2H2
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR2H2(Accession O95918), corresponding to amino acid residues S241-R291.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7932
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.