Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599949 Supplier Invitrogen™ Supplier No. PA599949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR2L2.

Odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2L2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR2L2
Gene Accession No. Q8NH16
Gene Alias HSHTPCRH07; HTPCRH07; Olfactory receptor 2L12; olfactory receptor 2L2; Olfactory receptor 2L4; olfactory receptor family 2 subfamily L member 2; olfactory receptor OR1-48 pseudogene; olfactory receptor, family 2, subfamily L, member 12; olfactory receptor, family 2, subfamily L, member 2; olfactory receptor, family 2, subfamily L, member 4 pseudogene; OR1-48; OR2L12; OR2L2; OR2L4P
Gene Symbols OR2L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR2L2(Accession Q8NH16), corresponding to amino acid residues E110-V160.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26246
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.