Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2L3/OR2L8 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5100983
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100983 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100983 Supplier Thermo Scientific Supplier No. PA5100983

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR2L3/2L8.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2L3/OR2L8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR2L8
Gene Accession No. Q8NG85, Q8NGY9
Gene Alias Olfactory receptor 2L3; olfactory receptor 2L8; olfactory receptor family 2 subfamily L member 3; olfactory receptor family 2 subfamily L member 8 (gene/pseudogene); olfactory receptor OR1-46; olfactory receptor, family 2, subfamily L, member 3; olfactory receptor, family 2, subfamily L, member 8; OR2L3; OR2L8
Gene Symbols OR2L3, OR2L8
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR2L3(Accession Q8NG85), corresponding to amino acid residues Q169-R219.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391190, 391192
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.