Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR2M4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5100985
Description
Antibody detects endogenous levels of total OR2M4.
Odorant receptor.
Specifications
OR2M4 | |
Polyclonal | |
Unconjugated | |
OR2M4 | |
HSHTPCRX18; HTPCRX18; novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor 2M4; olfactory receptor family 2 subfamily M member 4; olfactory receptor OR1-55; Olfactory receptor TPCR100; olfactory receptor, family 2, subfamily M, member 4; OR1-55; OR2M4; OST710; TPCR100 | |
Rabbit | |
Affinity chromatography | |
RUO | |
26245 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96R27 | |
OR2M4 | |
A synthesized peptide derived from human OR2M4(Accession Q96R27), corresponding to amino acid residues T240-Y290. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction