Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR2M4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2833140.1ML
Description
OR2M4 Polyclonal specifically detects OR2M4 in Human samples. It is validated for Western Blot.Specifications
OR2M4 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
26245 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
HSHTPCRX18, HTPCRX18Olfactory receptor TPCR100, novel 7 transmembrane receptor (rhodopsin family) protein, olfactory receptor 2M4, Olfactory receptor OR1-55, olfactory receptor, family 2, subfamily M, member 4, OST710OR1-55, TPCR100 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR2M4 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction