Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2T12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571176
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571176 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571176 Supplier Thermo Scientific Supplier No. PA571176
Add to Cart
Add to Cart

OR2T12 Polyclonal Antibody

OR2T12 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2T12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR2T12
Gene Accession No. Q8NG77
Gene Alias novel 7 transmembrane receptor (rhodopsin family) protein; olfactory receptor 2T12; olfactory receptor family 2 subfamily T member 12; olfactory receptor OR1-57; olfactory receptor, family 2, subfamily T, member 12; OR1-57; OR2T12
Gene Symbols OR2T12
Host Species Rabbit
Immunogen synthetic peptide directed towards the C-terminal region of human OR2T12.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 127064
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.