Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2T5/OR2T29 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103228
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103228 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103228 Supplier Thermo Scientific Supplier No. PA5103228

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR2T5/2T29.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2T5/OR2T29
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR2T5
Gene Accession No. Q6IEZ7, Q8NH02
Gene Alias fam; olfactory receptor 2T29; Olfactory receptor 2T5; olfactory receptor family 2 subfamily T member 29; olfactory receptor family 2 subfamily T member 5; olfactory receptor OR1-62; olfactory receptor, family 2, subfamily T, member 29; olfactory receptor, family 2, subfamily T, member 5; OR1-62; OR2T29; OR2T5
Gene Symbols OR2T29, OR2T5
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR2T29(Accession Q8NH02), corresponding to amino acid residues D74-Y124.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343563, 401993
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.