Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2V2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562106
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562106 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562106 Supplier Invitrogen™ Supplier No. PA562106
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WVNQSYTDGF FLLGIFSHST ADL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 70%.

Olfactory receptors interact with odorant molecules in thenose, to initiate a neuronal response that triggers the perceptionof a smell. The olfactory receptor proteins are members of a largefamily of G-protein-coupled receptors (GPCR) arising from singlecoding-exon genes. Olfactory receptors share a 7-transmembranedomain structure with many neurotransmitter and hormone receptorsand are responsible for the recognition and G protein-mediatedtransduction of odorant signals. The olfactory receptor gene familyis the largest in the genome. The nomenclature assigned to theolfactory receptor genes and proteins for this organism isindependent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2V2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration Lot-Specific
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OR2V2
Gene Accession No. Q96R30
Gene Alias olfactory receptor 2V2; Olfactory receptor 2V3; olfactory receptor family 2 subfamily V member 2; olfactory receptor OR5-3; olfactory receptor, family 2, subfamily V, member 2; olfactory receptor, family 2, subfamily V, member 3; OR2V2; OR2V3; OST713
Gene Symbols OR2V2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OR2V2. Recombinant protein control fragment (Product #RP-100794).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285659
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.