Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR2V2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575955
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575955 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575955 Supplier Invitrogen™ Supplier No. PA575955
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 96% (by SDS-PAGE).

Olfactory receptors interact with odorant molecules in thenose, to initiate a neuronal response that triggers the perceptionof a smell. The olfactory receptor proteins are members of a largefamily of G-protein-coupled receptors (GPCR) arising from singlecoding-exon genes. Olfactory receptors share a 7-transmembranedomain structure with many neurotransmitter and hormone receptorsand are responsible for the recognition and G protein-mediatedtransduction of odorant signals. The olfactory receptor gene familyis the largest in the genome. The nomenclature assigned to theolfactory receptor genes and proteins for this organism isindependent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR2V2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene OR2V2
Gene Accession No. Q96R30
Gene Alias olfactory receptor 2V2; Olfactory receptor 2V3; olfactory receptor family 2 subfamily V member 2; olfactory receptor OR5-3; olfactory receptor, family 2, subfamily V, member 2; olfactory receptor, family 2, subfamily V, member 3; OR2V2; OR2V3; OST713
Gene Symbols OR2V2
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 229-274 in Human OR2V2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 285659
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.