Learn More
Invitrogen™ OR2Z1 Polyclonal Antibody

Description
Immunogen sequence: MASDFLRGEG ATSYGGGAAQ I Highest antigen sequence identity to the following orthologs: Mouse - 52%, Rat - 57%.
Specifications
Specifications
| Antigen | OR2Z1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | OR2Z1 |
| Gene Accession No. | Q8NG97 |
| Gene Alias | Olfactory receptor 2Z1; olfactory receptor 2Z2; olfactory receptor family 2 subfamily Z member 1; olfactory receptor OR19-4; olfactory receptor, family 2, subfamily Z, member 1; olfactory receptor, family 2, subfamily Z, member 2; OR19-4; OR2Z1; OR2Z2 |
| Gene Symbols | OR2Z1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.