Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR3A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110062
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110062 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110062 Supplier Invitrogen™ Supplier No. PA5110062
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FYGSGIFNYM RLGSTKLSDK DKAVGIFNTV INPMLNPIIY SFRNPDVQSA IWRMLTGRRS LA.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR3A1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OR3A1
Gene Accession No. P47881
Gene Alias Olfactory receptor 17-40; olfactory receptor 3A1; olfactory receptor family 3 subfamily A member 1; olfactory receptor OR17-15; olfactory receptor, family 3, subfamily A, member 1; OLFRA03; OR17-40; OR17-82; OR3A1; OR40
Gene Symbols OR3A1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 254-315 of human OR3A1 (NP_002541.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4994
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.