Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR3A2/OR3A3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA5103229
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103229 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103229 Supplier Thermo Scientific Supplier No. PA5103229

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR3A2/3.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR3A2/OR3A3
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR3A2
Gene Accession No. P47888, P47893
Gene Alias Olfactory receptor 17-201; olfactory receptor 17-228; Olfactory receptor 3A2; olfactory receptor 3A3; Olfactory receptor 3A6; olfactory receptor 3A7; Olfactory receptor 3A8; olfactory receptor family 3 subfamily A member 2; olfactory receptor family 3 subfamily A member 3; olfactory receptor OR17-14; Olfactory receptor OR17-22; olfactory receptor, family 3, subfamily A, member 2; olfactory receptor, family 3, subfamily A, member 3; olfactory receptor, family 3, subfamily A, member 6; olfactory receptor, family 3, subfamily A, member 7; olfactory receptor, family 3, subfamily A, member 8 pseudogene; OLFRA04; OR17-137; OR17-14; OR17-16; OR17-201; OR17-228; OR228; OR3A2; OR3A3; OR3A6; OR3A7; OR3A8P
Gene Symbols OR3A2, OR3A3
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR3A3(Accession P47888), corresponding to amino acid residues G73-L123.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4995, 8392
Target Species Human, Non-human Primate, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.