Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR3A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116304
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116304 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116304 Supplier Invitrogen™ Supplier No. PA5116304
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OR3A3 Polyclonal Antibody for Western Blot

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR3A3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene OR3A3
Gene Accession No. P47888
Gene Alias olfactory receptor 17-201; Olfactory receptor 3A3; olfactory receptor 3A6; Olfactory receptor 3A7; olfactory receptor 3A8; olfactory receptor family 3 subfamily A member 3; olfactory receptor OR17-22; olfactory receptor, family 3, subfamily A, member 3; olfactory receptor, family 3, subfamily A, member 6; olfactory receptor, family 3, subfamily A, member 7; olfactory receptor, family 3, subfamily A, member 8 pseudogene; OR17-137; OR17-16; OR17-201; OR3A3; OR3A6; OR3A7; OR3A8P
Gene Symbols OR3A3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 255-281 amino acids from the C-terminal region of human OR3A3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8392
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.