Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR3A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599917
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599917 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599917 Supplier Invitrogen™ Supplier No. PA599917
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OR3A4.

Putative olfactory receptor 3A4 (OR3A4, OR3A4P) is an odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR3A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR3A4P
Gene Accession No. P47883
Gene Alias olfactory receptor 4A5; olfactory receptor family 4 subfamily A member 5; olfactory receptor OR11-111; olfactory receptor, family 4, subfamily A, member 5; OR11-111; OR4A5; seven transmembrane helix receptor
Gene Symbols OR3A4P
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR3A4P(Accession P47883), corresponding to amino acid residues G67-L117.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390756
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.