Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR4B1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575956
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575956 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575956 Supplier Invitrogen™ Supplier No. PA575956
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Olfactory receptors are G protein-coupled receptor proteins that localize to the cilia of olfactory sensory neurons where they display affinity for and bind to a variety of odor molecules. The genes encoding olfactory receptors comprise the largest family in the human genome. The binding of olfactory receptor proteins to odor molecules triggers a signal transduction cascade that leads to the production of cAMP via an olfactory-enriched adenylate cyclase. This event ultimately leads to transmission of action potentials to the brain and the subsequent perception of smell. OR4B1 (Olfactory receptor 4B1), also known as Olfactory receptor OR11-106, is a 309 amino acid multi-pass membrane protein that functions as an odorant receptor, effectively binding odor molecules and initiating the propagation of signals to the primary olfactory cortex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4B1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene OR4B1
Gene Accession No. Q8NGF8
Gene Alias olfactory receptor 4B1; olfactory receptor family 4 subfamily B member 1; olfactory receptor OR11-106; olfactory receptor, family 4, subfamily B, member 1; OR11-106; OR4B1; OST208
Gene Symbols OR4B1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 264-309 of Human OR4B1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 119765
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.