Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR4C45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546558 Supplier Invitrogen™ Supplier No. PA546558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: PTSFPTDKAV TVFCTLFTPM LNPLIYTLKN KEVKNVIKKL WKQIMTTDDK Sequence homology: Dog: 77%; Human: 100%; Rat: 77%.

OR4C45 is a odorant receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4C45
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OR4C45
Gene Accession No. A6NMZ5
Gene Alias olfactory receptor 4C45; olfactory receptor family 4 subfamily C member 45; olfactory receptor, family 4, subfamily C, member 45; OR4C45
Gene Symbols OR4C45
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human OR4C45 (aa 262-306).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403257
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.