Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR4C45 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100963
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100963 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100963 Supplier Thermo Scientific Supplier No. PA5100963
Add to Cart
Add to Cart

OR4C45 Polyclonal Antibody

OR4C45 Polyclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4C45
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR4C45
Gene Accession No. A6NMZ5
Gene Alias olfactory receptor 4C45; olfactory receptor family 4 subfamily C member 45; olfactory receptor, family 4, subfamily C, member 45; OR4C45
Gene Symbols OR4C45
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR4C45(Accession A6NMZ5), corresponding to amino acid residues G51-F101.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 403257
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.