Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR4D1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102193 Supplier Thermo Scientific Supplier No. PA5102193
Add to Cart
Add to Cart

OR4D1 Polyclonal Antibody

OR4D1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen OR4D1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OR4D1
Gene Accession No. Q15615
Gene Alias Olfactory receptor 4D1; olfactory receptor 4D3; olfactory receptor family 4 subfamily D member 1; olfactory receptor OR17-23 pseudogene; olfactory receptor TPCR16; olfactory receptor, family 4, subfamily D, member 1; olfactory receptor, family 4, subfamily D, member 3; olfactory receptor, family 4, subfamily D, member 4 pseudogene; OR17-23; OR4D1; OR4D3; OR4D4P; seven transmembrane helix receptor; TPCR16
Gene Symbols OR4D1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OR4D1(Accession Q15615), corresponding to amino acid residues K269-E310.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26689
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.