Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR4D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309782100UL
Description
OR4D1 Polyclonal specifically detects OR4D1 in Human samples. It is validated for Western Blot.Specifications
OR4D1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
olfactory receptor 4D1, Olfactory receptor 4D3, olfactory receptor OR17-23 pseudogene, Olfactory receptor TPCR16, olfactory receptor, family 4, subfamily D, member 1, olfactory receptor, family 4, subfamily D, member 3, olfactory receptor, family 4, subfamily D, member 4 pseudogene, OR17-23, OR4D3, OR4D4P, seven transmembrane helix receptor, TPCR16 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR4D1. Peptide sequence LLLISYTVILVMLRSHSGKARRKAASTCTTHIIVVSMIFIPCIYIYTWPF | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
26689 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction